Order Entry
Switzerland
ContactUsLinkComponent
4-Methyloctanoic acid for synthesis, Sigma-Aldrich®
4-Methyloctanoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.16104.0025
Supplier:  Merck
CAS Number:  
4-Methyloctanoic acid for synthesis, Sigma-Aldrich®
Catalog # 8.16104.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Molar Mass:
    158.24 g/mol
  • Boiling Point:
    292 - 293 °C
  • Flash Point:
    99 °C
  • Density:
    0.904 - 0.914 g/cm3 (20 °C)

Specification Test Results

About this item

Synonyms: 4-methylcaprylic acid, (R,S)-(+,-)-4-Methyloctanoic acid
 :
 :158.24 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ200
EINECS:259-404-2
UN:3265
 :GHS05
Storage Temperature:Ambient
CAS Number:54947-74-9
Formula:C₉H₁₈O₂
MDL Number:MFCD00051938