1-(4-Nitrophenylazo)-2-naphthol
Catalog # EHERC15875000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:293.28 g/mol
EINECS:229-093-8
CAS Number:6410-10-2
Formula:C₁₆H₁₁N₃O₃
MDL Number:MFCD00003908