


4-(Tributylstannyl)-1-tritylimidazole
Catalog # ACTIAS51346G1
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS06
:GHS08
:GHS09
CAS Number:208934-35-4
Formula:C₃₄H₄₄N₂Sn
MDL Number:MFCD10699150