ContactUsLinkComponent
Racks for EasyLoad® pipette tips
  GREI941310_10
Racks for EasyLoad® pipette tips
  GREI941310_10
  • Colour code:
    Yellow
  • Description:
    Box for EasyLoad® 200 tip insert
  • Pk:
    10
EasyLoad® is a simple, time- and space-saving refill system for pipette tips. The refill system also contains EasyLoad® racks specially developed for this purpose.
  • Autoclavable EasyLoad® racks
  • Sterile and non-sterile option
  • Racks are stackable
  • Colour-coded boxes
The modularly constructed EasyLoad® racks are available in different sizes. They consist of a polypropylene box with attached lid and a polypropylene insert plate. Users of the EasyLoad® refill system will find the insert plate included in the refill unit. The EasyLoad® racks are also particularly well suited for use with multi-channel pipettors, since the individual tips in the rack can be picked up with the same force.