Jars, white with red cap
Anbieter: Rosa Heinz GmbH
Zertifikate
Über diesen Artikel
PP, white with red PP cap.
- Excellent chemical resistance
- Excellent temperature resistance
Ideal for sampling, transportation and storage.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together