ContactUsLinkComponent
Pipette bulbs
Pipette bulbs
Catalog # 612-1939
Supplier:  DELTALAB
Pipette bulbs
Catalog # 612-1939
Supplier:  DELTALAB
Specifications
  • Colour:
    Translucent white
  • Description:
    Pipetting bulb, silicone, for glass and plastic pipettes (max. Ø 15 mm), 1,5 - 1,8 ml suction capacity, length 40 mm
  • Pk:
    50
Specifications
About this item
Pipetting bulb made of silicone (translucent white).
  • Designed for pipetting with plastic and glass pipettes
  • Silicone has a thermal and chemical resistance superior to latex