



Mercury(II) sulphate 200 g/l in diluted sulphuric acid for COD determination, Supelco®
Catalog # 1.04413.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Concentration (HgSO₄):198.0 - 202.0 g/l
- Concentration (H₂SO₄):173.0 - 180.0 g/l
- pH:0.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
About this item
:
:296.65 g/mol
Density:1,3 g/ml
ADR:8,II
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
UN:2992
:GHS06
:GHS08
:GHS05
:GHS09
CAS Number:7783-35-9
Formula:HgSO₄
MDL Number:MFCD00011047