Orders LinkContactUsLinkComponent
Heating tapes
Heating tapes
Supplier:   WINKLER
Heating tapes
Supplier:  WINKLER
Certificates
Heating tapes
25 Options Available Below
Product Details & Documents
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation
Product Family Options
Product Information
  • Availability
    Price
  • WINKWBS00200900050
    Heating tape, 0,9 m length, 50 W
    Heating tape, 0,9 m length, 50 W
    Log in for availability
    Log in for availability
    - SFr. 132.00
    SFr. 132.00
       SFr. 132.00
  • WINKWBS00200900100
    Heating tape, 0,9 m length, 100 W
    Heating tape, 0,9 m length, 100 W
    Log in for availability
    Log in for availability
  • WINKWBS00202900100
    Heating tape, 0,9 m length, 100 W
    Heating tape, 0,9 m length, 100 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00201100080
    Heating tape, 1,1 m length, 80 W
    Heating tape, 1,1 m length, 80 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00201800100
    Heating tape, 1,8 m length, 100 W
    Heating tape, 1,8 m length, 100 W
    Log in for availability
    Log in for availability
    - SFr. 207.00
    SFr. 207.00
       SFr. 207.00
  • WINKWBS00201800200
    Heating tape, 1,8 m length, 200 W
    Heating tape, 1,8 m length, 200 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00202200160
    Heating tape, 2,2 m length, 160 W
    Heating tape, 2,2 m length, 160 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00202700130
    Heating tape, 2,7 m length, 130 W
    Heating tape, 2,7 m length, 130 W
    Log in for availability
    Log in for availability
  • WINKWBS00202700270
    Heating tape, 2,7 m length, 270 W
    Heating tape, 2,7 m length, 270 W
    Log in for availability
    Log in for availability
  • WINKWBS00203100230
    Heating tape, 3,1 m length, 230 W
    Heating tape, 3,1 m length, 230 W
    Log in for availability
    Log in for availability
  • WINKWBS00204000200
    Heating tape, 4,0 m length, 200 W
    Heating tape, 4,0 m length, 200 W
    Log in for availability
    Log in for availability
    - SFr. 284.00
    SFr. 284.00
       SFr. 284.00
  • WINKWBS00204000410
    Heating tape, 4,0 m length, 410 W
    Heating tape, 4,0 m length, 410 W
    Log in for availability
    Log in for availability
  • WINKWBS00204600350
    Heating tape, 4,6 m length, 350 W
    Heating tape, 4,6 m length, 350 W
    Log in for availability
    Log in for availability
  • WINKWBS00206300320
    Heating tape, 6,3 m length, 320 W
    Heating tape, 6,3 m length, 320 W
    Log in for availability
    Log in for availability
  • WINKWBS00206300640
    Heating tape, 6,3 m length, 640 W
    Heating tape, 6,3 m length, 640 W
    Log in for availability
    Log in for availability
  • WINKWBS00207300550
    Heating tape, 7,3 m length, 550 W
    Heating tape, 7,3 m length, 550 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00209500470
    Heating tape, 9,5 m length, 470 W
    Heating tape, 9,5 m length, 470 W
    Log in for availability
    Log in for availability
  • WINKWBS00209500950
    Heating tape, 9,5 m length, 950 W
    Heating tape, 9,5 m length, 950 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00211000820
    Heating tape, 11,0 m length, 820 W
    Heating tape, 11,0 m length, 820 W
    Log in for availability
    Log in for availability
  • WINKWBS00213500670
    Heating tape, 13,5 m length, 670 W
    Heating tape, 13,5 m length, 670 W
    Log in for availability
    Log in for availability
  • WINKWBS00213501340
    Heating tape, 13,5 m length, 1340 W
    Heating tape, 13,5 m length, 1340 W
    Log in for availability
    Log in for availability
  • WINKWBS00215501160
    Heating tape, 15,5 m length, 1160 W
    Heating tape, 15,5 m length, 1160 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00216000780
    Heating tape, 16,0 m length, 780 W
    Heating tape, 16,0 m length, 780 W
    Log in for availability
    Log in for availability
  • WINKWBS00216001570
    Heating tape, 16,0 m length, 1570 W
    Heating tape, 16,0 m length, 1570 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • WINKWBS00218301370
    Heating tape, 18,3 m length, 1370 W
    Heating tape, 18,3 m length, 1370 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST