Orders LinkContactUsLinkComponent
Microtest manifolds
Microtest manifolds
Supplier:   DRUMMOND
Microtest manifolds
Supplier:  DRUMMOND
Certificates
Microtest manifolds
11 Options Available Below
Product Details & Documents
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
Product Family Options
Product Information
  • Availability
    Price
  • DRUM3-000-090
    Microtest manifold, SS/PTFE, straight, 4-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, straight, 4-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 601.00
    SFr. 601.00
    1 * 1 ST
       SFr. 601.00
  • DRUM3-000-097
    Microtest manifold, SS/PTFE, straight, 4-place for 24 well plate, 20 mm centre-to-centre, pin length 17,5 mm
    Microtest manifold, SS/PTFE, straight, 4-place for 24 well plate, 20 mm centre-to-centre, pin length 17,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 731.00
    SFr. 731.00
    1 * 1 ST
       SFr. 731.00
  • DRUM3-000-093
    Microtest manifold, SS/PTFE, straight, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, straight, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 731.00
    SFr. 731.00
    1 * 1 ST
       SFr. 731.00
  • DRUM3-000-096
    Microtest manifold, SS/PTFE, straight, 12-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, straight, 12-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 954.00
    SFr. 954.00
    1 * 1 ST
       SFr. 954.00
  • DRUM3-000-102
    Microtest manifold, SS/PTFE, straight, 16-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, straight, 16-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’060.00
    SFr. 1’060.00
    1 * 1 ST
       SFr. 1’060.00
  • DRUM3-000-101
    Microtest manifold, SS/PTFE, straight, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, straight, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’270.00
    SFr. 1’270.00
    1 * 1 ST
       SFr. 1’270.00
  • DRUM3-000-098
    Microtest manifold, SS/PTFE, bent, 4-place for 24 well plate, 20 mm centre-to-centre, pin length 17,5 mm
    Microtest manifold, SS/PTFE, bent, 4-place for 24 well plate, 20 mm centre-to-centre, pin length 17,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 731.00
    SFr. 731.00
    1 * 1 ST
       SFr. 731.00
  • DRUM3-000-094
    Microtest manifold, SS/PTFE, bent, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, bent, 8-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 731.00
    SFr. 731.00
    1 * 1 ST
       SFr. 731.00
  • DRUM3-000-099
    Microtest manifold, SS/PTFE, bent, 12-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, bent, 12-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 954.00
    SFr. 954.00
    1 * 1 ST
       SFr. 954.00
  • DRUM3-000-104
    Microtest manifold, SS/PTFE, bent, 16-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, bent, 16-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’060.00
    SFr. 1’060.00
    1 * 1 ST
       SFr. 1’060.00
  • DRUM3-000-103
    Microtest manifold, SS/PTFE, bent, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Microtest manifold, SS/PTFE, bent, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’270.00
    SFr. 1’270.00
    1 * 1 ST
       SFr. 1’270.00