HPLC columns, Acclaim™ Explosives E2
Acclaim™ Explosives E2 columns are used to resolve the nitroaromatics and nitramines, the novel and unique chemistries of these columns provide superior resolution with complementary selectivities.
- Simple isocratic elution conditions
- Rugged column with good lot-to-lot reproducibility
Frequently Bought Together