Screw caps with valves, Mininert
Supplier: Thermo Fisher Scientific
Certificates
About this item
Mininert valves are screw caps that have integrated resealable valves to allow repeated and unlimited syringe-needle access to samples eliminates the septum-boring that occurs with repeated puncture of traditional septa. Made of chemical-resistant PTFE coating, they provide an inert, leak-tight, high-pressure seal.
Frequently Bought Together