Order Entry
Switzerland
ContactUsLinkComponent
Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Katalognr. BDAA354010
Anbieter:  Corning
   Low Manufacturing Impact
CAS-Nummer:  
Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Katalognr. BDAA354010
Anbieter:  Corning
   Low Manufacturing Impact
CAS-Nummer:  

Technische Daten

  • Pk:
    100 µG
  • Protein/peptide type:
    Native
  • Species:
    Mouse
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein/peptide name:
    Epidermal Growth Factor

Technische Daten

Über diesen Artikel

Epidermal growth factor (EGF) is a low-molecular weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro. EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

  • Receptor Grade

EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

Mouse natural EGF are tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. Human recombinant EGF are shown to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. All products are filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.