ClipTip™ Racks for Tips
Supplier: Thermo Fisher Scientific
ClipTip™ Racks for Tips
Supplier: Thermo Fisher Scientific
Certificates
About this item
Utilize Thermo Scientific™ ClipTip™ Pipette tip Racks, available in three sizes.
- Small rack: For use with ClipTip™ 20 and 50 Pipette tips
- Medium rack: For use with ClipTip™ 200 and 300 Pipette tips
- Large rack: For use with ClipTip™ 1000 Pipette tips
Ordering information: Racks are supplied without tips and inserts.
4 Options Available Below
Limited Time Sale
Select items below are currently on sale
Product Details & Documents
Utilize Thermo Scientific™ ClipTip™ Pipette tip Racks, available in three sizes.
- Small rack: For use with ClipTip™ 20 and 50 Pipette tips
- Medium rack: For use with ClipTip™ 200 and 300 Pipette tips
- Large rack: For use with ClipTip™ 1000 Pipette tips
Ordering information: Racks are supplied without tips and inserts.
Specifications for Product Family
Specifications
- Catalog No:
- LABS94410619
- 613-2226
- 613-2221
- 613-2227
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- LABS94410619Empty rack, colour code orange (for ClipTip 300 extended length)Empty rack, colour code orange (for ClipTip 300 extended length)Log in for availability1 * 8 STSFr. 41.90Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LABS94410619Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-2226Empty rack, large, colour code blue (for ClipTip 1000)Empty rack, large, colour code blue (for ClipTip 1000)Log in for availabilityLog in for availability1 * 8 ST1 * 8 ST - SFr. 22.00SFr. 22.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 8 STSFr. 22.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2226Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-2221Empty rack, medium, colour code yellow and orange (for ClipTip 200 and 300)Empty rack, medium, colour code yellow and orange (for ClipTip 200 and 300)Log in for availabilityLog in for availability1 * 10 ST1 * 10 ST - SFr. 22.00SFr. 22.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 10 STSFr. 22.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2221Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-2227Empty rack, small, colour code pink and purple (for ClipTip 20 and 50)Empty rack, small, colour code pink and purple (for ClipTip 20 and 50)Log in for availabilityLog in for availability1 * 10 ST1 * 10 ST - SFr. 22.00SFr. 22.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 10 STSFr. 22.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-2227Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact