Round cover glasses, Nunc Thermanox
Supplier: Thermo Fisher Scientific
Certificates
About this item
These cover glasses remain flat during normal use.
- Resistant to common solvents
- Autofluorescent from 380 to 545 nm
- Surfaced treatment on one side for optimal cell attachment and growth
Specifications
- Catalog No:
- 734-2117
- 734-2173
- 734-2118
- 734-2172
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement