ContactUsLinkComponent
Alcohol-by-volume (ABV) kit, Eclipse
Alcohol-by-volume (ABV) kit, Eclipse
Catalog # BELL44-839
Supplier:  BELLINGHAM STANLEY
undefined
Alcohol-by-volume (ABV) kit, Eclipse
Catalog # BELL44-839
Supplier:  BELLINGHAM STANLEY
Specifications
  • Division:
    0,5
  • Pk:
    1
  • Range:
    0,98 - 1,05 SG
Specifications
About this item
This alcohol-by-volume (ABV) kit combines a refractometer and a hydrometer for the determination of alcohol content of finished beers wine and cider as well as for monitoring ferment in bottle beers and sparkling wines.
  • Quick and easy process to carry out
  • Special hydrometer scale covers most samples
  • Internet calculator via PC or Web enabled mobile device
The alcohol content of a beer, wine and cider may easily be established by combining the results of two simple test measurements, that of a refractometer (RI-Zeiss) and a hydrometer (SG). This kit is able to achieve an alcohol by volume accuracy of 0,5% ABV.
Certifications: Calibration showing traceability to ICUMSA and NIST.
Ordering information: Each components needs to be ordered separately to for a ABV kit.
Delivery: Supplied with paper lookup table for the calculation of results.
Caution: Due to the achievable accuracy of this measurement system, results may not be suitable for duty or tax declaration purposes.
Handheld refractometer
Product Information