ContactUsLinkComponent
Barrier tips, GEN3™, Axygen®
Barrier tips, GEN3™, Axygen®
Catalog # 732-7515
Supplier:  Corning
undefined
Barrier tips, GEN3™, Axygen®
Catalog # 732-7515
Supplier:  Corning
Specifications
  • Packed:
    50 racks, 100 each
  • Sterile:
    +
  • Volume:
    100 - 1000 µl
  • Seq:
    04
  • Tip style:
    Standard
  • Pk:
    5.000
Specifications
About this item
These multi-barrier filter pipette tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact assuring the valuable samples and research have not been compromised.
  • Effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material
  • Proprietary filter design prevents possible PCR inhibition
  • Maxymum Recovery® technology for low sample retention and high yield
  • Hinged rack for convenience and ease of use
  • Certified DNase-/RNase-free and non pyrogenic
  • Certified human gDNA and PCR inhibition free