Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pipettes
Pipette Controllers
Accessories for pipette controller
Accessories for pipette controller
Accessories for pipette controller
Catalog # BELC1225-88050
Supplier:
BELLCO GLASS
Expand image
Accessories for pipette controller
Catalog # BELC1225-88050
Supplier:
BELLCO GLASS
Specifications
Description:
Tissue culture nose piece, retrofit kit
Pk:
1
Specifications
About this item
Product Details
Product Details