Pipette controller, Pipet-Aid
Portable Pipet-Aid, 230 V / 50 Hz
BELC1225-85220
Marketsource
Bellco Pipet-Aids are lightweight, compact units that are ideal for use under laminar flow hoods.
- Rechargeable, self-contained units
- Two-finger volume control provides precise fluid uptake and dispensing
- Three independent speed ranges can be set for both fill and dispense rates
Basic Pipet-Aid: 230 V pump with optional tissue culture nose piece and/or air filter.
Portable Pipet-Aid: Rechargeable self-contained unit with removable stand.
: Portable pipet-aid is supplied with battery charger and tissue culture nose piece.
- Description:Portable Pipet-Aid, 230 V / 50 Hz
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



