Order Entry
Switzerland
ContactUsLinkComponent
D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade for biotechnology
D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade for biotechnology
Catalog # E543-100ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade for biotechnology
Catalog # E543-100ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Clarity:
    Pass

Specification Test Results

About this item

CAS Number:57-50-1
 :
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
 :342.3 g/mol
EINECS:200-334-9