Order Entry
Switzerland
ContactUsLinkComponent
Hydrochloric acid 5%, ARISTAR® for calibration blank in ICP
Corrosive
Hydrochloric acid 5%, ARISTAR® for calibration blank in ICP
Catalog # 456494P
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Hydrochloric acid 5%, ARISTAR® for calibration blank in ICP
Catalog # 456494P
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
ADR:8,III
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324