Order Entry
Switzerland
ContactUsLinkComponent
Lactic acid standardised solution 1.0 N in aqueous solution
Corrosive
Lactic acid standardised solution 1.0 N in aqueous solution
Catalog # 35628.AP
CAS Number:  
Corrosive
Lactic acid standardised solution 1.0 N in aqueous solution
Catalog # 35628.AP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

About this item

 :
 :90.08 g/mol
EINECS:200-018-0
 :GHS05
Storage Temperature:Ambient
CAS Number:50-21-5
 :598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520