Order Entry
Switzerland
ContactUsLinkComponent
Tetraethylammonium hydroxide ∼1 mol/l in aqueous solution, Supelco®
Corrosive
Tetraethylammonium hydroxide ∼1 mol/l in aqueous solution, Supelco®
Catalog # 86636-250ML
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Tetraethylammonium hydroxide ∼1 mol/l in aqueous solution, Supelco®
Catalog # 86636-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :147.26 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
EINECS:201-073-3
UN:3267
 :GHS05
CAS Number:77-98-5
Formula:C₈H₂₁NO
MDL Number:MFCD00009024