Methyl-4-hydroxypicolinate hydrochloride
Catalog # ACTIAS23233G1
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:1256633-27-8
Formula:C₇H₈ClNO₃
MDL Number:MFCD18071002