Surface cleaning and disinfectant products, Pursept® AF
: Brand
A disinfecting detergent for cleaning and disinfection of floors, walls and fittings in one operation.
- Appropriate for use in the food sector
- Clear, green liquid with fresh smell
- Mildly alkaline of 1% solution with pH value of approximately 10
As recommended by the RKI, Bundesgesundheitsblatt 01/2004.
Frequently Bought Together