Metallurgical microscopes, BA310 MET
: MOTIC
These metallurgical microscopes are designed for viewing opaque materials such as minerals and metals.
- High quality glass objectives enabling improved contrast and image quality
- Flexible focus dedicated to large samples
- Various lighting options for incident and/or transmitted light
Suitable for educational environments of engineering or material professionals.
NOTE: For increased illumination, replace the halogen bulb by an LED module.
- Catalog No:
- MOTI1100100402501
- MOTI1100100402521
- 630-2690
- 630-2689
- MOTI1101001704512
- MOTI1101001704402
- MOTI1100100402511
- MOTI1101001704611
- 630-2044
- MOTI1101001704601
- MOTI1101000201982
- 630-2605
- MOTI1101001901609
- MOTI1101001704392
- Head:
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- Eyepieces:
- WF 10×/20 mm
- WF 10×/20 mm
- WF 10×/20 mm
- WF 10×/20 mm
- WF 10×/20 mm
- WF 10×/20 mm
- Nosepiece:
- Quintuple, reversed
- Quintuple, reversed
- Quintuple, reversed
- Quintuple, reversed
- Quintuple, reversed
- Quintuple, reversed
- Focusing system:
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Illumination:
- Halogen
- Halogen
- Halogen
- Halogen
- Halogen
- Halogen