Orders LinkContactUsLinkComponent
Welding face shield
Welding face shield
Welding shield, helmet-mounted, EU version, Welding protection
Catalog # CENUS903NCE
Welding face shield
Catalog # CENUS903NCE
Welding face shield
Welding face shield
Welding shield, helmet-mounted, EU version, Welding protection
Price
SFr. 69.60
Log in for availability
1 * 1 ST
Product Details & Documents
This lightweight helmet-mounted welding shield is highly impact resistant.
  • Shield is made of robust Polyamide PA6 (Nylon)
  • Supplied with Polycarbonate chipping lens
  • Compatible with all Centurion helmets except for the Spartan helmet
Window: 110×90 mm
Certifications: EN 175
Specifications
  • Type:
    Welding protection
  • Description:
    Welding shield, helmet-mounted, EU version