Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Boron standard solution, 5000 mg/L B in water
VWR®, Boron standard solution, 5000 mg/L B in water
Catalog # 88380.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Boron standard solution, 5000 mg/L B in water
Catalog # 88380.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    1.000 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item

This certified reference material is produced using a high-purity starting material, acid from sub-boiling and 18 MOhm deionised water.