Order Entry
Switzerland
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid) 0.01 mol/l in aqueous solution
EDTA (ethylenediamine tetraacetic acid) 0.01 mol/l in aqueous solution
Catalog # HACH2700653
Supplier:  Hach
CAS Number:  
undefined
EDTA (ethylenediamine tetraacetic acid) 0.01 mol/l in aqueous solution
Catalog # HACH2700653
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :292.25 g/mol
Storage Temperature:Ambient
EINECS:200-449-4
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541