Order Entry
Switzerland
ContactUsLinkComponent
 
Potassium nitrate 10% in aqueous solution
  REGEEFSKNO
 :  REAGECON
 :  
Potassium nitrate 10% in aqueous solution
  REGEEFSKNO
 :  REAGECON
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

 :
 :101.1 g/mol
 :Ambient
 :231-818-8
 :7757-79-1
 :KNO₃
 :MFCD00011409