Order Entry
Switzerland
ContactUsLinkComponent
1,3-Propanediol
1,3-Propanediol
Catalog # XXXX5495260000
Supplier:  Chemicals
CAS Number:  
1,3-Propanediol
Catalog # XXXX5495260000
Supplier:  Chemicals
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241

Specifications

About this item

 :
 :76.1 g/mol
Merck Index:12,09845
Boiling Pt:214 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.0597 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-997-3
CAS Number:504-63-2
Formula:HO(CH₂)₃OH
MDL Number:MFCD00002949