ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Corrosive
Sodium hydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Certificates
Specification Test Results
  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective