Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenolphthalein
Phenolphthalein 10 g/l in ethanol
Phenolphthalein 10 g/l in ethanol
:
PROLABO CMP
Phenolphthalein 10 g/l in ethanol
:
PROLABO CMP
:
77-09-8
:
:
C₂₀H₁₄O₄
:
MFCD00005913
:
318.33 g/mol
:
∼78 °C (1013 hPa)
:
∼0.85 g/cm³ (20 °C)
:
Ambient
:
201-004-7
:
1170
:
3,II
:
GHS02
:
GHS08
Frequently Bought Together
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Test strips, nitrate/nitrite, Dosatest
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Test strips for free chlorine, MQuant®, Supelco®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
: Merck
1,5-Diphenylcarbazide ≥97.0% analytical reagent
: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
: Merck