Order Entry
Switzerland
ContactUsLinkComponent
Potassium hydroxide 8.0 M (8.0 N) in aqueous solution volumetric solution, Fluka™
Potassium hydroxide 8.0 M (8.0 N) in aqueous solution volumetric solution, Fluka™
  P4494-50ML
 :  
Potassium hydroxide 8.0 M (8.0 N) in aqueous solution volumetric solution, Fluka™
  P4494-50ML
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370
  • Environmentally Preferable:

 

 

  • Assay titr.:
    7.92 - 8.08 mol/l
  • Clearness of the solution:
    complying

 

 

 : Potash caustic, Caustic potash
 :
 :56.11 g/mol
 :112 °C (1.013 hPa)
 :-6 °C
 :–6 °C
 :1.510 g/cm3 (20 °C)
 :Ambient
 :215-181-3
 :1814
 :8,II
 :GHS05
 :GHS07
 :1310-58-3
 :KOH
 :MFCD00003553