Order Entry
Switzerland
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid)
Irritant
EDTA (ethylenediamine tetraacetic acid)
Catalog # PIER17892
CAS Number:  
undefined
Irritant
EDTA (ethylenediamine tetraacetic acid)
Catalog # PIER17892
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
 :GHS07
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541