Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Certificates
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

3 Options Available Below

Product Details & Documents

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236358

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J62449.K2
  • J62449.K3
  • J62449.K7
  • Appearance:
  • Colourless
  • Colourless
  • Colourless
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • MDL No.:
  • MFCD00236358
  • MFCD00236358
  • MFCD00236358

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • J62449.K2
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
    Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 99.20
    SFr. 99.20
    1 * 1 L
       SFr. 99.20

    Specifications:
    More
    Specifications
    Cat. No.
    J62449.K2
    Appearance
    Colourless
    Physical form
    Liquid
    MDL No.
    MFCD00236358
  • J62449.K3
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
    Glass bottle
    2 L
    Log in for availability
    Log in for availability
    1 * 2 L
    1 * 2 L - SFr. 149.00
    SFr. 149.00
    1 * 2 L
       SFr. 149.00

    Specifications:
    More
    Specifications
    Cat. No.
    J62449.K3
    Appearance
    Colourless
    Physical form
    Liquid
    MDL No.
    MFCD00236358
  • J62449.K7
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
    Powder packages
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - SFr. 269.00
    SFr. 269.00
    1 * 4 L
       SFr. 269.00

    Specifications:
    More
    Specifications
    Cat. No.
    J62449.K7
    Appearance
    Colourless
    Physical form
    Liquid
    MDL No.
    MFCD00236358
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.