Methyl-4-chloro-7-methoxyquinoline-6-carboxylate
Catalog # ACTIAS12683G1
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.6700 g/mol
CAS Number:205448-66-4
Formula:C₁₂H₁₀ClNO₃
MDL Number:MFCD17016133