

5-Benzylmercaptotetrazole (BMT, BTT) 0.3 mol/l in acetonitrile activator reagent, Novabiochem®
Supplier: Merck
Certificates
About this item
CAS Number:21871-47-6
:
Formula:C₈H₈N₄S
MDL Number:MFCD00068731
:192.24 g/mol
UN:1648
ADR:3,II
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together