Buffer, EDTA buffer solution pH 8,0 (0,25 mol/L)
EDTA buffer solution pH 8,0 (0,25 mol/l), 1 KIT
Catalog # EURLEMR254001BK
Supplier: EUROCLONE
Buffer, EDTA buffer solution pH 8,0 (0,25 mol/L)
Catalog # EURLEMR254001BK
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 KIT
- Description:EDTA buffer solution pH 8,0 (0,25 mol/l)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



