ContactUsLinkComponent
Cleanroom shirts, 7R011
undefined
Cleanroom shirts, 7R011
  • Catalog No:
  • KWIN1006355402XL
  • KWIN1006355403XL
  • KWIN100635540L
  • KWIN100635540M
  • KWIN100635540S
  • KWIN100635540XL
  • KWIN100635540XS
  • Model:
  • 7R011, XA32
  • 7R011, XA32
  • 7R011, XA32
  • 7R011, XA32
  • 7R011, XA32
  • 7R011, XA32
  • 7R011, XA32
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Fabric:
  • Polyester/carbon
  • Polyester/carbon
  • Polyester/carbon
  • Polyester/carbon
  • Polyester/carbon
  • Polyester/carbon
  • Polyester/carbon
  • Weight:
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
Frequently Bought Together