ContactUsLinkComponent
Sealing film, self-adhesive for fluorescence measurement
Sealing film, self-adhesive for fluorescence measurement
Codice catalogo 738-3001
Nome del fornitore:  Brand
undefined
Sealing film, self-adhesive for fluorescence measurement
Codice catalogo 738-3001
Nome del fornitore:  Brand
Specifiche
  • Optical property:
    Light blocking
  • Description:
    Sealing film, self-adhesive, for fluorescence measurement, vinyl, black, single sheets
  • Pk:
    50
Specifiche
Informazioni su questo articolo
Black sealing film made of vinyl is highly suitable for fluorescence measurements because of its light-absorbing characteristics. Temperature range: −40 to +80 °C.