About this item
These Hei-VAP devices, with hand lift, are ideally suited for all standard applications. Parameters can be controlled with the simple turn of a knob.
- Large dial controls for adjustment of rotation speed and bath temperature
- Required angle and height of the evaporating flask can adjusted individually
- Patented EasyClip flask clamp allows the simple and safe removal of the evaporating flask due to the force of unscrewing the Easy-Clip
- Detachable operating panel enables safe and convenient handling from outside closed fume hoods
- Separate on/off switch for heating prevents unintentional heat-up – the button is illuminated for visual control
- FDA-compliant, transparent and UV resistant tubing, with higher chemical resistance
210 °C safety heating bath, with handles and pour spout, will power off if temperature overshoots by 5 °C or runs dry. The bath power cable coupling complies with protection class IP 67 to prevent corrosion and short circuits.
Glassware set G1
Diagonal condenser for all standard distillations; option with the most affordable price.
Glassware set G3
Vertical condenser for all standard distillations; option with the smallest space requirements, also available as XL version (condensing surface of 2200 cm² for up to 40% higher evaporation rates).
Glassware set G5
Dry ice condenser for low boiling solvents.
Glassware set G6
Vertical condenser; centrepiece with valve for reflux distillation.
As standard, models are supplied with a detachable control panel with a 1,3 m cable. Therefore parameters can be changed outside of a closed fume hood, providing greater operator safety and ultimate process control. An optional remote control operating panel allows the user to step away from the evaporator and still monitor all process data increasing flexibility at work.
Please note: A vacuum source, vacuum-check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.