ContactUsLinkComponent
BOD seed inoculum, POLYSEED
BOD seed inoculum, POLYSEED
Catalog # HACH2918700
Supplier:  Hach
CAS Number:  
undefined
BOD seed inoculum, POLYSEED
Catalog # HACH2918700
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    50
  • Description:
    BOD seed inoculum, POLYSEED
Specifications
About this item
BOD seed inoculum for use in biochemical oxygen demand (BOD5) test procedures. A blend of specialised microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.
Ordering information: Package of 50 capsules. International Laboratory Supply (InterLab), LTD.