ContactUsLinkComponent
Lauryl tryptose (with MUG) broth tubes, concentrated
Lauryl tryptose (with MUG) broth tubes, concentrated
Catalog # HACH2182115
Supplier:  Hach
CAS Number:  
undefined
Lauryl tryptose (with MUG) broth tubes, concentrated
Catalog # HACH2182115
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    15
  • Standards:
    -
  • Description:
    Lauryl tryptose (with MUG) broth tubes
Specifications
About this item
Lauryl tryptose broth with MUG provides a selective medium which is used for enumeration of E.coli and other coliform bacteria in water using the Most Probable Number (MPN) method.
  • Save time and labour costs with ready to use MPN tubes, with minimal training requirements
  • Regulatory-accepted methods for applications in drinking water, wastewater, ambient water, food and beverage