Orders LinkContactUsLinkComponent
Turbidity standards, Stablcal® | Avantor
Turbidity standards, Stablcal®
 :   Hach
Turbidity standards, Stablcal®
 :  Hach
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results
Dokumente
Product Family Options
  • Description
    Turbidity
    Pk
  • Stablcal® stabilised formazine standard
    0,06 NTU
    100 ml
    1 * 100 mL
    1 * 100 mL - SFr. 193.00
    SFr. 193.00
    1 * 100 mL
       SFr. 193.00
  • Stablcal® stabilised formazine standard
    0,10 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    0,10 NTU
    1 L
    1 * 1 L
    1 * 1 L - SFr. 461.00
    SFr. 461.00
    1 * 1 L
       SFr. 461.00
  • Stablcal® stabilised formazine standard
    <0,1 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 113.00
    SFr. 113.00
    1 * 100 mL
       SFr. 113.00
  • Stablcal® stabilised formazine standard
    <0,1 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 186.00
    SFr. 186.00
    1 * 500 mL
       SFr. 186.00
  • Stablcal® stabilised formazine standard
    <0,1 NTU
    1 L
    1 * 1 L
    1 * 1 L - SFr. 393.00
    SFr. 393.00
    1 * 1 L
       SFr. 393.00
  • Stablcal® stabilised formazine standard
    0,30 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    0,30 NTU
    1 L
    1 * 1 L
    1 * 1 L - SFr. 402.00
    SFr. 402.00
    1 * 1 L
       SFr. 402.00
  • Stablcal® stabilised formazine standard
    0,50 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    1 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    1 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
  • Stablcal® reference suspension I acc. Ph. EUR 6.0
    3 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 410.00
    SFr. 410.00
    1 * 100 mL
       SFr. 410.00
  • Stablcal reference suspension II acc. Ph. EUR 6.0
    6 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 410.00
    SFr. 410.00
    1 * 100 mL
       SFr. 410.00
  • Stablcal® stabilised formazine standard
    10 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    10 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
  • Stablcal reference suspension III acc. Ph. EUR 6.0
    18 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 410.00
    SFr. 410.00
    1 * 100 mL
       SFr. 410.00
  • Stablcal® stabilised formazine standard
    20 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    20 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
  • Stablcal® stabilised formazine standard
    20 NTU
    1 L
    1 * 1 L
    1 * 1 L - SFr. 459.00
    SFr. 459.00
    1 * 1 L
       SFr. 459.00
  • Stablcal reference suspension IV acc. Ph. EUR 6.0
    30 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 410.00
    SFr. 410.00
    1 * 100 mL
       SFr. 410.00
  • Stablcal® stabilised formazine standard
    40 NTU
    1 L
    1 * 1 L
    1 * 1 L - SFr. 415.00
    SFr. 415.00
    1 * 1 L
       SFr. 415.00
  • Stablcal® stabilised formazine standard
    100 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    100 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
  • Stablcal® stabilised formazine standard
    200 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    200 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
  • Stablcal® stabilised formazine standard
    800 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    800 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
  • Stablcal® stabilised formazine standard
    1,0 NTU
    1 L
    1 * 1 L
    1 * 1 L - SFr. 459.00
    SFr. 459.00
    1 * 1 L
       SFr. 459.00
  • Stablcal® stabilised formazine standard
    1000 NTU
    100 mL
    1 * 100 mL
    1 * 100 mL - SFr. 194.00
    SFr. 194.00
    1 * 100 mL
       SFr. 194.00
  • Stablcal® stabilised formazine standard
    1000 NTU
    500 mL
    1 * 500 mL
    1 * 500 mL - SFr. 375.00
    SFr. 375.00
    1 * 500 mL
       SFr. 375.00
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..