ContactUsLinkComponent
Laboratory glass cleaner, borer 12 BASIC
Laboratory glass cleaner, borer 12 BASIC
Catalog # DECO500325.00-K05W
Supplier:  Borer Chemie
undefined
Laboratory glass cleaner, borer 12 BASIC
Catalog # DECO500325.00-K05W
Supplier:  Borer Chemie
Specifications
  • Description:
    borer 12 BASIC
  • Seq:
    02
  • Pk:
    7 kg
Specifications
About this item
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
  • Rinseable without residues and guarantees therefore consistent and error-free results
  • Perfect results even at low concentrations
  • High cleaning performance due to excellent swelling and dispersing capabilities
  • Working solution suitable for multiple use with a broad cleaning spectrum
  • Ideal as an adequate substitute for chromosulphuric acid, not toxic and environmentally friendly
  • EDTA-, NTA- and chlorine-free