m-Endo broth
: Hach
m-Endo broth detects total coliform bacteria (colonies with a greenish-gold sheen). For potable water samples, confirm with Lauryl Tryptose and brilliant green bile broth tubes. Samples that test positive for total coliform bacteria should be further analysed for the presence of faecal coliform or E. coli using EC Medium or EC Medium with MUG respectively.
- Save valuable time and money using prepared media for membrane filtration methods
Frequently Bought Together