Order Entry
Switzerland
ContactUsLinkComponent
Rotors for MIKRO 200 centrifuge
Rotors for MIKRO 200 centrifuge
Catalog # HETT2434
Supplier:  Hettich
   Low Manufacturing Impact
Rotors for MIKRO 200 centrifuge
Catalog # HETT2434
Supplier:  Hettich
   Low Manufacturing Impact

Specifications

  • Description:
    Fixed angle rotor 2434, 24×2,0 ml, 15000 min-1, 21382 ×g
  • Seq:
    05
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Pk:
    1
  • Environmental certification:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)

Specifications

About this item