PEEK-encased frits
Supplier: VICI JOUR RESEARCH
Certificates
About this item
Made of stainless steel or titanium. Titanium frits are resistant to corrosion, biocompatible and recommended for protein analysis.
- Intended for use with HPLC columns or in-line filters
- Eliminate poorly swept volume and improves peak symmetry
The compressed PEEK ring forms a seal between the bottom of the fitting and the column end, preventing the mobile phase and the sample from entering the previously accessible poorly swept areas.
Frequently Bought Together