Flow-through filters
These flow-through filters are made of PTFE. The filter membrane can be replaced easily by unscrewing the sample funnel. Plastic tubing can be connected with olives.
- Suitable for overpressure or vacuum up to about 5 bar
- Matching adapter for 2× tube sizes included
- Anti-adhesive surface allows easy cleaning
: Delivery without filter membranes, please order separately.
- Catalog No:
- ROTHY937.1
- ROTHY935.1
- Filters:
- Ø 90 mm
- Ø 25 mm
- Filtration area:
- 52,0 cm²
- 3,1 cm²
- Max. temperature:
- 121 °C
- 121 °C
Frequently Bought Together