Order Entry
Switzerland
ContactUsLinkComponent
 
VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 22 Elements in Nitric Acid 5% for ICP
Catalog # 84790.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 22 Elements in Nitric Acid 5% for ICP
Catalog # 84790.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard in 5% dilute nitric acid, ARISTAR® for ICP

Specifications

About this item

22 elements 100 mg/L: As, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.